missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IFN-gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 1710.00 €
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | 88915 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18726738
![]() |
R&D Systems
MAB585-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18784708
![]() |
R&D Systems
MAB585-100 |
100 μg |
429.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18786338
![]() |
R&D Systems
MAB585-500 |
500 μg |
1710.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Rat samples. It is validated for Western Blot, Neutralization.Specifications
| IFN-gamma | |
| Western Blot 1 ug/mL, Neutralization 0.5-1.5 ug/mL | |
| Monoclonal | |
| Mouse | |
| Rat | |
| P01581 | |
| 3458 | |
| E. coli-derived recombinant rat IFN-gamma Gln23-Cys156 Accession # P01581 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat IFN-gamma in Western blots. In Western blots, this antibody shows less than 1% cross-reactivity with recombinant mouse (rm) IFN-gamma and no cross-reactivity with rhIFN-gamma . |
| 88915 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNG | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title