missing translation for 'onlineSavingsMsg'
Learn More

Rat IgG2a Isotype Control (KLH/G2a-1-1) [Janelia Fluor 669], Novus Biologicals™

Product Code. 31846118 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31846118 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31846118 Supplier Novus Biologicals Supplier No. NBP151104JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

IgG2a Monoclonal antibody specifically detects IgG2a in Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Control
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Control
Classification Monoclonal
Clone KLH/G2a-1-1
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen KLH
Quantity 0.25 mL
Regulatory Status RUO
Target Species Rat
Content And Storage Store at 4°C in the dark.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.