missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
209.00 € - 1705.00 €
Specifications
| Antigen | IL-4 |
|---|---|
| Clone | 56567 |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721668
![]() |
R&D Systems
MAB504-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18759567
![]() |
R&D Systems
MAB504-100 |
100 μg |
429.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713027
![]() |
R&D Systems
MAB504-500 |
500 μg |
1705.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-4 Monoclonal specifically detects IL-4 in Rat samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
| IL-4 | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Rat | |
| P20096.2 | |
| 3565 | |
| E. coli-derived recombinant rat IL-4 Cys25-Ser147 Accession # P20096.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat IL-4 in ELISAs and Western blots. Does not cross-react with recombinant human IL-4 or recombinant mouse IL-4. |
| 56567 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| IL4 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title