missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IL-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB506R-100
This item is not returnable.
View return policy
Description
IL-6 Monoclonal specifically detects IL-6 in Rat samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| IL-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A | |
| E. coli-derived recombinant rat IL-6, Phe25-Thr211, Accession No. P20607 | |
| 100 μg | |
| 3569.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 53325R | |
| ELISA Capture (Matched Antibody Pair) 2-8 μg;/mL | |
| P20607 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects rat IL-6 in direct ELISAs. | |
| Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction