missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Rat Set

Product Code. 10695003
Change view
Click to view available options
Quantity:
20 x 3 nmol
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10695003 20 x 3 nmol Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10695003 Supplier Ambion™ Supplier No. 1330003
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Set

Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
TRUSTED_SUSTAINABILITY

Specifications

Description Stealth RNAi™ Pre-designed siRNA Set, 20 x 3nmol, Rat Set Target, Good Knockdown, Low Off-target Effects, Standard Purity, Used For Treating Parkinson's Disease
Final Product Type Stealth RNAi™ Pre-designed siRNA Set
For Use With (Application) For treating Parkinson's disease
Product Type Stealth RNAi™ Pre-designed siRNA Set
Purity Standard purity
Quantity 20 x 3 nmol
Sample Size 20 x 3nmol
Target Rat Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.