Learn More
Rat TfR (Transferrin R) Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB9294R-100UG
 View more versions of this product
Product Code. 18723338
  • 491.00 € / 100µg
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TfR (Transferrin R)
Monoclonal
Alexa Fluor 647
Supplied in a saline solution containing BSA and Sodium Azide.
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7037
Rat
IgG2a
Flow Cytometry
OX26
Flow Cytometry 0.5 ug/10^6 cells
CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
PHA activated rat lymphocytes
100 μg
Primary
Detects rat TfR (Transferrin R) in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.