missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat TNF-alpha Antibody, R&D Systems™
Recombinant Monoclonal Antibody
Brand: R&D Systems MAB510R-01M
This item is not returnable.
View return policy
Description
TNF-alpha Monoclonal specifically detects TNF-alpha in Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| TNF-alpha | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7124 | |
| Rat | |
| Purified |
| ELISA, Neutralization | |
| 45418R | |
| Neutralization 10-40 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P16599 | |
| Tnf | |
| E. coli-derived recombinant rat TNF-alpha Accession No. P16599 | |
| 1 mg | |
| Primary | |
| Detects rat TNF-alpha in direct ELISAs. In ELISAs, this antibody shows less than 3% cross-reactivity with recombinant mouse (rm) TNF-alpha and less than 0.2% cross-reactivity with rhTNF-alpha , rpTNF-alpha , and rhTNF-beta . | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction