missing translation for 'onlineSavingsMsg'
Learn More

Scientific Industries Ratcheting Clamp, Vortex-Genie Max

Product Code. 17281762 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie Max
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17281762 Vortex-Genie Max Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17281762 Supplier Scientific Industries Supplier No. SIMX050
  • ONLINE EXCLUSIVE PRICE
    120.75 €
    102.40 € / Pack of 1
    Save 18.35 €
    15% Off
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ratcheting Clamp, Vortex-Genie Max is an extra ratcheting clamp to add additional rows for the Vortex-Genie Max.

  • Extra ratcheting clamp for additional rows on Genie Temp-shakers
TRUSTED_SUSTAINABILITY

Specifications

Product Type Ratcheting Clamp
Color Black
For Use With (Equipment) Vortex-Genie Max
Packaging Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.