Learn More
Invitrogen™ RAVER2 Recombinant Rabbit Monoclonal Antibody (24GB810)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557679
 View more versions of this product
Product Code. 30275555
  • 198.00 € / 20µL
Description
RAVER2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
RAVER2 may bind single-stranded nucleic acids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RAVER2
Recombinant Monoclonal
Unconjugated
RAVER2
A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2
Rabbit
Affinity Chromatography
RUO
55225
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB810
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9HCJ3
RAVER2
A synthesized peptide derived from human RAVER2 (aa 481-691).
20 μL