missing translation for 'onlineSavingsMsg'
Learn More

Raxatrigine hydrochloride, MedChemExpress

Product Code. 16461547
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461547 50 mg 50mg
16445358 10 mg 10mg
16435986 100 mg 100mg
16418077 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16461547 Supplier Medchem Express Supplier No. HY12796A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Raxatrigine hydrochloride (GSK-1014802 hydrochloride) is a novel small molecule state-dependent sodium channel blocker; Nav1.7 sodium channel inhibitor.

Raxatrigine hydrochloride (GSK-1014802 hydrochloride) is a novel small molecule state-dependent sodium channel blocker

Specifications

Chemical Name or Material Raxatrigine hydrochloride
CAS 934240-31-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H20ClFN2O2
Quantity 50 mg
Synonym GSK-1014802 hydrochlorideCNV1014802 hydrochloride
Solubility Information DMSO : ≥ 31 mg/mL (88.36 mM) ∣H2O : 14.29 mg/mL (40.73 mM; Need ultrasonic)
SMILES O=C([C@H]1N[C@@H](C2=CC=C(OCC3=CC=CC=C3F)C=C2)CC1)N.[H]Cl
Molecular Weight (g/mol) 350.82
Formula Weight 350.82
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.