missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RB1 Antibody (XZ91), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-30762-25ug
This item is not returnable.
View return policy
Description
RB1 Monoclonal antibody specifically detects RB1 in Chicken,Human,Mink,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RB1 | |
| Monoclonal | |
| Unconjugated | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| A recombinant protein corresponding to amino acids 387 to 928 of the human retinoblastoma sequence | |
| 25 μg | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 5925 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| XZ91 | |
| Phosphate buffered saline | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Chicken, Human, Mink, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction