Learn More
Novus Biologicals™ RB1 Antibody (XZ91), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-30762-100ug
 View more versions of this product
Product Code. 30067235
  • 508.00 € / 100µg
Description
RB1 Monoclonal antibody specifically detects RB1 in Chicken,Human,Mink,Mouse,Rat samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RB1
Monoclonal
Unconjugated
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
A recombinant protein corresponding to amino acids 387 to 928 of the human retinoblastoma sequence
100 μg
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
5925
Store at -20°C. Avoid freeze/thaw cycles.
IgG1
Immunoprecipitation, Western Blot
XZ91
Phosphate buffered saline
Mouse
Protein A or G purified
RUO
Primary
Chicken, Human, Mink, Mouse, Rat
Purified