missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1CC1 Mouse, anti-Human, Clone: 1G8C8, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | RB1CC1 |
|---|---|
| Clone | 1G8C8 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18039435
|
Proteintech
68564-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18036695
|
Proteintech
68564-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RB1CC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RB1CC1 | |
| 1000 μg/mL | |
| Liquid | |
| Human, Mouse, Rat | |
| CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC | |
| Primary |
| 1G8C8 | |
| Western Blot, ELISA | |
| Mouse | |
| 9821 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title