Learn More
RB1CC1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL750-17250-100UL
 View more versions of this product
Product Code. 18094735
  • 450.00 € / 100µL
Description
The RB1CC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 750.
Specifications
Specifications
RB1CC1
1000 μg/mL
CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC
100 μL
9821
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Flow Cytometry (Intracellular Staining)
CoraLite Plus 750
Rabbit
Primary
Human, Mouse, Rat
150 kDa