missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RbAp46 Antibody (2A10C2), Novus Biologicals™
Mouse Monoclonal Antibody
287.00 € - 511.00 €
Specifications
| Antigen | RbAp46 |
|---|---|
| Clone | 2A10C2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 |
| Applications | Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30024776
|
Novus Biologicals™
NBP3-27151-0.025ml |
0.025 mL |
287.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30034165
|
Novus Biologicals™
NBP3-27151-0.1ml |
0.1 mL |
511.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RbAp46 Monoclonal antibody specifically detects RbAp46 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ ImmunofluorescenceSpecifications
| RbAp46 | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors | |
| PBS | |
| 5931 | |
| IgG1 | |
| Protein G purified |
| 2A10C2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| G1/S transition control protein-binding protein RbAp46, Histone acetyltransferase type B subunit 2, histone-binding protein RBBP7, MGC138867, Nucleosome-remodeling factor subunit RBAP46, RbAp46, RBBP-7, retinoblastoma binding protein 7, Retinoblastoma-binding protein 7MGC138868, Retinoblastoma-binding protein p46, retinoblastoma-binding protein RbAp46 | |
| Purified recombinant fragment of human RbAp46 (AA: 1-200) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title