Learn More
RBCK1 Antibody (CL4289) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59048UV
Product Code. 30293248
  • 737.00 € / 0.10mL
Description
RBCK1 Monoclonal antibody specifically detects RBCK1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBCK1
Monoclonal
DyLight 350
C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4
This antibody was developed using a recombinant protein derived from Q9BYM8, with the exact immunogen sequence remaining proprietary.
0.1 mL
Zinc Finger
10616
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL4289
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified