missing translation for 'onlineSavingsMsg'
Learn More

RBCK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-27105

 View more versions of this product

Product Code. 18770274

  • 361.00 € / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

RBCK1 Polyclonal specifically detects RBCK1 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RBCK1
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4
Rabbit
Protein A purified
RUO
Primary
The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, and chimpanzee, 94% in rhesus monkey, 89% in new world monkey, and macaque, 83% in cow, guinea pig, and mole rat, 78% in dog, elephant, and rabbit, 72% in panda, 67% in opossum, 61% in rat, and 55% homologous in mouse.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
1.0 mg/mL
Western Blot 2-4ug/ml
Q9BYM8
RBCK1
A portion of amino acids 150-200 from human RBCk1 was used as the immunogen for this antibody.
0.1 mg
Zinc Finger
10616
Human, Mouse, Primate
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only