missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody (1B7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92693AF647
This item is not returnable.
View return policy
Description
RBFOX3/NeuN Monoclonal specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
| RBFOX3/NeuN | |
| Monoclonal | |
| Alexa Fluor 647 | |
| A6NFN3 | |
| RBFOX3 | |
| N-terminal 99 amino acids of human FOX3 as expressed in and purified from E. coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| 146713 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1B7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Porcine, Mouse, Rat and other Mammals. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction