missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
234.00 € - 513.00 €
Specifications
| Antigen | RBFOX3/NeuN |
|---|---|
| Concentration | 1 mg/ml |
| Dilution | Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18035847
|
Novus Biologicals
NBP1-92716SS |
0.025 mL |
234.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18766783
|
Novus Biologicals
NBP1-92716 |
0.1 mL |
513.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBFOX3/NeuN Polyclonal specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.Specifications
| RBFOX3/NeuN | |
| Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating 1:500-1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| A6NFN3 | |
| 146713 | |
| The N-terminal 100 amino acids of human RBFOX3/NeuN as expressed in and purified from E. coli. [UniProt# A6NFN3] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 48 kDa |
| 1 mg/ml | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3 | |
| RBFOX3 | |
| IgG | |
| Affinity purified | |
| Human, bovine, porcine, mouse, rat and other mammals. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title