missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM26 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89007
This item is not returnable.
View return policy
Description
RBM26 Polyclonal antibody specifically detects RBM26 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
| RBM26 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| acidic rich RS domain containing 2, ARRS2, C13orf10, chromosome 13 open reading frame 10, CTCL tumor antigen se70-2, cutaneous T-cell lymphoma tumor antigen se70-2, FLJ20957, MGC133295, MGC133296, PRO1777, RNA binding motif protein 26, RNA-binding motif protein 26, RNA-binding protein 26, RP11-255E21.1, SE70-2, ZC3H17 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: PPLTPLQPSGMDAPPNSATSSVPTVVTTGIHHQPPPAPPSLFTADTYDTDGY | |
| 0.1 mL | |
| Cell Biology | |
| 64062 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction