Learn More
RBM3 Antibody (CL0296) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52906MFV500
Product Code. 30287454
  • 759.00 € / 0.10mL
Description
RBM3 Monoclonal antibody specifically detects RBM3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBM3
Monoclonal
mFluor Violet 500 SE
IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL
This antibody was developed using a recombinant protein derived from P98179, with the exact immunogen sequence remaining proprietary.
0.1 mL
metabolism
5935
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0296
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified