missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SDEHVLEELE TEGERQLKSL LQHQLDTSVS IEECMSKKES FAPGTMYKPF GKEAAGTMTL SQFQTLHEKD QETASLRELG LNETEILIWK SHVSGEKKTK LRATPEAIQN RLQDIEERIS ERQRILCLPQ RFAKSKQLTR REMEI Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 93%.
RBM41, known as RNA Binding Motif Protein 41, is identified as a novel component of the minor spliceosome, particularly interacting with its paralog, U11/U12-65K. Both RBM41 and U11/U12-65K share similar C-terminal RNA recognition motifs (RRMs), which are crucial for their role in spliceosomal activity. Distinctly, RBM41 has been associated with spliceosomal functions, although its precise cellular functions remain largely unexplored. Understanding the structural and functional parallels between RBM41 and U11/U12-65K aids in elucidating the mechanisms underlying minor spliceosome activity. The minor spliceosome itself is involved in the splicing of U12-type introns, thus playing a significant role in the regulation of gene expression. Additionally, RBM41's functional characterization in protein complexes and its involvement with RNA-binding activities enrich our understanding of gene regulation and cellular processes where the spliceosome plays a pivotal role. Research on RBM41 contributes to broader insights into RNA splicing and molecular genetics.
Specifications
Specifications
| Antigen | RBM41 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RBM41 |
| Gene Accession No. | Q96IZ5 |
| Gene Alias | BB131279; D330023I21Rik; Rbm41; RNA binding motif protein 41; RNA-binding motif protein 41; RNA-binding protein 41 |
| Gene Symbols | RBM41 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
