missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RBMX Recombinant Rabbit Monoclonal Antibody (K01_1S36)

Product Code. 30830408
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830408 50 μL
30827674 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830408 Supplier Invitrogen™ Supplier No. MA564942
 Restricted Item
Estimated Shipment: 22-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RVGRQERGLP PSME.

hnRNP G is a glycoprotein, which was originally identified as an autoantigen from German shepherd dogs with lupus-like syndrome. The gene encoding hnRNP G is located on chromosome Xq26 and is ubiquitously expressed. It contains one RNP-consensus RNA binding domain (RBD) and is related to RBMY, which is involved in spermatogenesis, and hnRNP G-T, which is a testis specific protein. All three proteins interact with Tra2b and therefore, are involved in pre-mRNA splicing.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen RBMX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1S36
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene RBMX
Gene Accession No. P38159
Gene Alias glycoprotein p43; Heterogeneous nuclear ribonucleoprotein G; hnRNP G; HNRNPG; hnRNP-G; Hnrpg; MRXS11; RBMX; Rbmxp1; RBMXRT; RNA binding motif protein, X chromosome; RNA binding motif protein, X-linked; RNA-binding motif protein, X chromosome; RNA-binding motif protein, X chromosome retrogene; RNA-binding motif protein, X chromosome retrogene-like; RNA-binding motif protein, X chromosome, N-terminally processed; RNMX
Gene Symbols RBMX
Host Species Rabbit
Immunogen A synthetic peptide of human hnRNP G (aa 341-354).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27316
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.