missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBPJL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24664-1-AP-150UL
This item is not returnable.
View return policy
Description
In mouse, recombining binding protein L (RBP-L) is a transcription factor that binds to DNA sequences almost identical to that bound by the Notch receptor signaling pathway transcription factor RBP-J. However, unlike RBP-J, RBP-L does not interact with Notch receptors. RBP-L has been shown to activate transcription in concert with Epstein-Barr virus nuclear antigen-2 (EBNA2). The protein encoded by this gene is similar in sequence to the mouse RPB-L protein and Drosophila suppressor of hairless protein.Specifications
| RBPJL | |
| Polyclonal | |
| Unconjugated | |
| RBPJL | |
| RBP L, RBPJL, RBPL, RBPSUHL, SUH, SUHL, Transcription factor RBP L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11317 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UBG7 | |
| RBPJL | |
| RBPJL Fusion Protein Ag20284 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction