missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RBPSUHL 293T Cell Transient Overexpression Lysate

Product Code. 18322539 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18322539 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322539 Supplier R&D Systems™ Supplier No. H00011317T02
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Overexpression Lysate

TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze-thaw cycles.
Product Line Novus
Target RBPSUHL
Synonym RBPL, RBP-L, RBPSUHL, recombination signal binding protein for immunoglobulin kappa J region-like, recombining binding protein suppressor of hairless (Drosophila)-like, recombining binding protein suppressor of hairless-like protein, SUH, SUHL, Transcription factor RBP-L
Gene Symbol RBPJL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.