missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBX1 Antibody (SR1116), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21835-50ul
This item is not returnable.
View return policy
Description
RBX1 Monoclonal antibody specifically detects RBX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| RBX1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1116 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| BA554C12.1, EC 6.3.2.-, FLJ60363, MGC13357, Protein ZYP, Rbx1, Regulator of cullins 1, RING box protein 1, RING finger protein 75, ring-box 1, ring-box 1, E3 ubiquitin protein ligase, RING-box protein 1, RNF75MGC1481, ROC1E3 ubiquitin-protein ligase RBX1, ZYP protein | |
| A synthesized peptide derived from human RBX1 (Uniprot #: P62877) | |
| 50 μg | |
| Cancer, Cell Cycle and Replication, Hypoxia, Neuroscience, Wnt Signaling Pathway, Zinc Finger | |
| 9978 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction