missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RC74 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | RC74 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607701
|
Novus Biologicals
NBP2-93568-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623760
|
Novus Biologicals
NBP2-93568-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RC74 Polyclonal antibody specifically detects RC74 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| RC74 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| CPSF2L, FLJ10871, INT9, integrator complex subunit 9, Protein related to CPSF subunits of 74 kDa, RC74, RC-74 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 459-658 of human INTS9 (NP_060720.2). KLLKEVQPLHVVCPEQYTQPPPAQSHRMDLMIDCQPPAMSYRRAEVLALPFKRRYEKIEIMPELADSLVPMEIKPGISLATVSAVLHTKDNKHLLQPPPRPAQPTSGKKRKRVSDDVPDCKVLKPLLSGSIPVEQFVQTLEKHGFSDIKVEDTAKGHIVLLQEAETLIQIEEDSTHIICDNDEMLRVRLRDLVLKFLQKF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55756 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title