missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCC2 Polyclonal Antibody

Description
Peptide sequence: RIRSLACGKS SIIVAADEST ISWGPSPTFG ELGYGDHKPK SSTAAQEVKT Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Required for completion of mitosis and cytokinesis. May function as a guanine nucleotide exchange factor for the small GTPase RAC1.
Specifications
Specifications
| Antigen | RCC2 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | RCC2 |
| Gene Accession No. | Q8BK67, Q9P258 |
| Gene Alias | 2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; protein RCC2; Protein RCC2 homolog; RCC1-like; RCC1-like protein TD-60; RCC2; regulator of chromosome condensation 2; RGD1309986; Td60; td-60; Telophase disk protein of 60 kDa; wu:fb36a03; zgc:77115 |
| Gene Symbols | RCC2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?