missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RCE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140156
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LTDPKLYGSL PLCVLLERAG DSEAPLCS Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.
This gene encodes an integral membrane protein which is classified as a member of the metalloproteinase family. This enzyme is thought to function in the maintenance and processing of CAAX-type prenylated proteins.
Specifications
| RCE1 | |
| Polyclonal | |
| Unconjugated | |
| RCE1 | |
| CAAX prenyl protease 2; CAAX prenyl protein protease RCE1; D19Ertd283e; D19Ertd98e; FACE2; FACE-2; farnesylated protein-converting enzyme 2; Farnesylated proteins-converting enzyme 2; hRCE1; prenyl protein peptidase RCE1; prenyl protein-specific endoprotease 2; Ras and a-factor-converting enzyme 1 homolog; Ras converting CAAX endopeptidase 1; Rce1; RCE1 homolog; RCE1 homolog, prenyl protein peptidase; RCE1 homolog, prenyl protein peptidase (S. cerevisiae); RCE1 homolog, prenyl protein protease; RCE1A; RCE1B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9986 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y256 | |
| RCE1 | |
| Recombinant protein corresponding to Human RCE1. Recombinant protein control fragment (Product #RP-109390). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction