missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RDH12 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15868303
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15868303 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15868303 Supplier Invitrogen™ Supplier No. PA569896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 85%; Dog: 92%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Pig: 86%; Rabbit: 85%; Rat: 79%; Yeast: 79%; Zebrafish: 92%.

The protein encoded by this gene is an NADPH-dependent retinal reductase whose highest activity is toward 9-cis and all-trans-retinol. The encoded enzyme also plays a role in the metabolism of short-chain aldehydes but does not exhibit steroid dehydrogenase activity. Defects in this gene are a cause of Leber congenital amaurosis type 3 (LCA3).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RDH12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RDH12
Gene Accession No. Q96NR8
Gene Alias A930033N07Rik; All-trans and 9-cis retinol dehydrogenase; double substrate specificity short-chain dehydrogenase/reductase 2; double substrate-specificity short chain dehydrogenase/reductase 2; DSSDR2; ds-SDR2; LCA13; LCA3; Rdh12; Retinol dehydrogenase 12; retinol dehydrogenase 12 (all-trans and 9-cis); retinol dehydrogenase 12 (all-trans/9-cis/11-cis); retinol dehydrogenase 12, all-trans and 9-cis; RP53; SDR7C2; short chain dehydrogenase/reductase family 7C member 2; short chain dehydrogenase/reductase family 7C, member 2; wu:fj43a10; zgc:92430
Gene Symbols RDH12
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human RDH12.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 145226
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.