missing translation for 'onlineSavingsMsg'
Learn More

Reboxetine mesylate, MedChemExpress

Product Code. 16456098
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456098 10 mg 10mg
16448697 5 mg 5mg
16442237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16456098 Supplier Medchem Express Supplier No. HY14560C
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Reboxetine mesylate (FCE20124 mesylate) is a potent, selective, and specific noradrenaline reuptake inhibitor (NARI) for the research of depression. Reboxetine mesylate inhibits the uptake of norepinephrine, with a Ki of 8 nM.

Specifications

Chemical Name or Material Reboxetine mesylate
CAS 98769-84-7
Color White
Purity Grade Notes Research
Molecular Formula C20H27NO6S
Quantity 10 mg
Synonym FCE20124 mesylate PNU155950E mesylate
Solubility Information DMSO : 125 mg/mL (305.25 mM; Need ultrasonic) ∣H2O : 50 mg/mL (122.10 mM; Need ultrasonic)
SMILES CS(=O)(O)=O.CCOC1=C(C=CC=C1)O[C@]([C@@]2([H])CNCCO2)([H])C3=CC=CC=C3.[relative stereochemistry]
Molecular Weight (g/mol) 409.5
Formula Weight 409.5
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.