Learn More
Macherey-Nagel™ Receiver Plate
Product Code. 11942542
Change view
Click to view available options
Pore Size:
20 μm
50 μm
Includes:
Filter Frits
Hydrophilized Filter Frits
Unit Size:
Pack of 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Includes unitSize
11942542 50 μm Filter Frits Pack of 4
11902542 20 μm Hydrophilized Filter Frits Pack of 4
11932542 20 μm Filter Frits Pack of 4
11912542 50 μm Hydrophilized Filter Frits Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11942542 Supplier Macherey-Nagel™ Supplier No. 740688.4
Only null left
Add to basket
Add to basket
Use for filtration in 96-well format. Macherey Nagel™ Receiver Plates are important accessories for high throughput work.
  • Receiver plate accessory
  • High-throughput applications
  • Filter frits
  • Suitable for centrifugation and vacuum
Specifications
Pore Size 50 μm
Includes Filter Frits