missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Fibritin Protein

A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Fibritin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7036-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017140

  • 3590.00 € / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_049771.1
Fibritin Protein
Research Use Only
wac
Recombinant Protein
E. coli
TDIVLNDLPF VDGPPAEGQS RISWIKNGEE ILGADTQYGS EGSMNRPTVS VLRNVEVLDK NIGILKTSLE TANSDIKTIQ GILDVSGDIE ALAQIGINKK DISDLKTLTS EHTEILNGTN NTVDSILADI GPFNAEANSV YRTIRNDLLW IKRELGQYTG QDINGLPVVG NPSSGMKHRI INNTDVITSQ GIRLSELETK FIESDVGSLT IEVGNLREEL GPKPPSFSQN VYSRLNEIDT KQTTVESDIS AIKTSIGYPG NNSIITSVNT NTDNIASINL ELNQSGGIKQ RLTVIETSIG SDDIPSSIKG QIKDNTTSIE SLNGIVGENT SSGLRANVSW LNQIVGTDSS GGQPSPPGSL LNRVSTIETS VSGLNNAVQN LQVEIGNNSA GIKGQVVALN TLVNGTNPNG STVEERGLTN SIKANETNIA SVTQEVNTAK GNISSLQGDV QALQEAGYIP EAPRDGQAYV RKDGEWVFLS TFLSPA
Greater than 90% as determined by SDS-PAGE.
1258630
1 mg
Not determined.
Full Length
Bacteriophage
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers