missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bacteriophage Fibritin Protein
A DNA sequence encoding the Enterobacteria phage T4 (Bacteriophage T4) Fibritin, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | NP_049771.1 |
---|---|
Gene ID (Entrez) | 1258630 |
Name | Fibritin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007140
|
enQuireBio™
QP7036-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047140
|
enQuireBio™
QP7036-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097130
|
enQuireBio™
QP7036-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027140
|
enQuireBio™
QP7036-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037140
|
enQuireBio™
QP7036-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16017140
|
enQuireBio™
QP7036-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_049771.1 | |
Fibritin Protein | |
Not determined. | |
Full Length | |
Bacteriophage | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1258630 | |
Research Use Only | |
wac | |
Recombinant Protein | |
E. coli | |
TDIVLNDLPF VDGPPAEGQS RISWIKNGEE ILGADTQYGS EGSMNRPTVS VLRNVEVLDK NIGILKTSLE TANSDIKTIQ GILDVSGDIE ALAQIGINKK DISDLKTLTS EHTEILNGTN NTVDSILADI GPFNAEANSV YRTIRNDLLW IKRELGQYTG QDINGLPVVG NPSSGMKHRI INNTDVITSQ GIRLSELETK FIESDVGSLT IEVGNLREEL GPKPPSFSQN VYSRLNEIDT KQTTVESDIS AIKTSIGYPG NNSIITSVNT NTDNIASINL ELNQSGGIKQ RLTVIETSIG SDDIPSSIKG QIKDNTTSIE SLNGIVGENT SSGLRANVSW LNQIVGTDSS GGQPSPPGSL LNRVSTIETS VSGLNNAVQN LQVEIGNNSA GIKGQVVALN TLVNGTNPNG STVEERGLTN SIKANETNIA SVTQEVNTAK GNISSLQGDV QALQEAGYIP EAPRDGQAYV RKDGEWVFLS TFLSPA | |
Greater than 90% as determined by SDS-PAGE. |