missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine ApoAI Protein
A DNA sequence encoding the Bos taurus (Bovine) ApoAI, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8986-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_776667.2 | |
ApoAI Protein | |
Research Use Only | |
APOA1 | |
Recombinant Protein | |
Yeast | |
DDPQSSWDRV KDFATVYVEA IKDSGRDYVA QFEASALGKQ LNLKLLDNWD TLASTLSKVR EQLGPVTQEF WDNLEKETAS LRQEMHKDLE EVKQKVQPYL DEFQKKWHEE VEIYRQKVAP LGEEFREGAR QKVQELQDKL SPLAQELRDR ARAHVETLRQ QLAPYSDDLR QRLTARLEAL KEGGGSLAEY HAKASEQLKA LGEKAKPVLE DLRQGLLPVL ESLKVSILAA IDEASKKLNA Q | |
Greater than 90% as determined by SDS-PAGE. |
281631 | |
500 μg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |