missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Bovine Enteropeptidase/Enterokinase His (C-Term) Protein

Product Code. 18678576 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 IU
250 IU
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18678576 100 IU Each
18648058 250 IU Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18678576 Supplier Novus Biologicals™ Supplier No. NBP261360100IU

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Enzyme Activity
Formulation 50 mM Tris-HCl, pH 8.0, 0.5 M NaCl
Gene ID (Entrez) 5651
Molecular Weight (g/mol) MolecularWeight-theroretical: 28 kDa
Quantity 100 IU
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of Enteropeptidase/Enterokinase, His as determined by LAL method.
Gene Alias EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15
Gene Symbol TMPRSS15
Research Category Epitope Tags, Immunology, Innate Immunity
Purity or Quality Grade Peptide affinity purified
Protein Enteropeptidase/Enterokinase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.