missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. elegans SNAP25 Protein
A cDNA sequence encoding the SNAP25 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13532-1mg
Additional Details : Weight : 0.01000kg
Specifications
SNAP25 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGDDDIPEG LEAINLKMNA TTDDSLESTR RMLALCEESK EAGIKTLVML DDQGEQLERCEGALDTINQD MKEAEDHLKG MEKCCGLCVL PWNKTDDFEK TEFAKAWKKD DDGGVISDQPRITVGDSSMG PQGGYITKIT NDAREDEMDE NVQQVSTMVG NLRNMAIDMS TEVSNQNRQL DRIHDKAQSN EVRVESANKR AKNLITK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
C. elegans | |
Untagged | |
The protein contains 20mM Tris-HCl pH 7.5, 50mM NaCl, 5mM DTT, 1mM EDTA and 10% Glycerol. |