missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. elegans SNAP25 Protein
A cDNA sequence encoding the SNAP25 was constructed and used to recombinantly synthesize the protein.
131.00 € - 2610.00 €
Specifications
Name | SNAP25 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | C. elegans |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913879
|
enQuireBio™
QP13532-10UG |
10 μg |
131.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933879
|
enQuireBio™
QP13532-50UG |
50 μg |
203.00 €
50µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923879
|
enQuireBio™
QP13532-1MG |
1 mg |
2610.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
SNAP25 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
C. elegans | |
Untagged | |
The protein contains 20mM Tris-HCl pH 7.5, 50mM NaCl, 5mM DTT, 1mM EDTA and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSGDDDIPEG LEAINLKMNA TTDDSLESTR RMLALCEESK EAGIKTLVML DDQGEQLERCEGALDTINQD MKEAEDHLKG MEKCCGLCVL PWNKTDDFEK TEFAKAWKKD DDGGVISDQPRITVGDSSMG PQGGYITKIT NDAREDEMDE NVQQVSTMVG NLRNMAIDMS TEVSNQNRQL DRIHDKAQSN EVRVESANKR AKNLITK | |
Greater than 95.0% as determined by SDS-PAGE. |