missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant C. elegans SNAP25 Protein

A cDNA sequence encoding the SNAP25 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13532-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15933879

  • 203.00 € / 50µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SNAP25 Protein
Research Use Only
Recombinant Protein
E. coli
MSGDDDIPEG LEAINLKMNA TTDDSLESTR RMLALCEESK EAGIKTLVML DDQGEQLERCEGALDTINQD MKEAEDHLKG MEKCCGLCVL PWNKTDDFEK TEFAKAWKKD DDGGVISDQPRITVGDSSMG PQGGYITKIT NDAREDEMDE NVQQVSTMVG NLRNMAIDMS TEVSNQNRQL DRIHDKAQSN EVRVESANKR AKNLITK
Greater than 95.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
C. elegans
Untagged
The protein contains 20mM Tris-HCl pH 7.5, 50mM NaCl, 5mM DTT, 1mM EDTA and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers