missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Canine IL-6 Protein

Product Code. p-7111058 Shop All Bio Techne Products
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
This item is not returnable. View return policy

Product Code. 18282283

Brand: Novus Biologicals™ NBP25956450UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids21-207 of Canine IL-6 The Recombinant Canine IL-6 Protein is derived from Baculovirus. The Recombinant Canine IL-6 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH 7.4) containing 10% glycerol
Gene ID (Entrez) 3569
Molecular Weight (g/mol) 22 kDa
Name Recombinant Canine IL-6 Protein
Purification Method >90%, by SDS-PAGE
Quantity 50 μg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias B cell stimulatory factor-2, B-cell differentiation factor, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFN-beta-2, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF
Gene Symbol IL6
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Canine
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.