missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog CMA1 / Chymase 1 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) CMA1 / Chymase 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5843-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001013442.1 | |
CMA1 / Chymase 1 Protein | |
Research Use Only | |
CMA1 | |
Recombinant Protein | |
E. coli | |
IIGGTESKPH SRPYMAHLEI LTLRNHLASC GGFLIRRNFV LTAAHCAGRF IMVTLGAHNI QKKEDTWQKL EVIKQFPHPK YDDLTLRHDI MLLKLKEKAN LTLAVGTLPL SPQFNFVPPG RMCRVAGWGK RQVNGSGSDT LQEVKLRLMD PQACRHYMAF DHNLQLCVGN PRKTKSAFKG DSGGPLLCAG VAQGIVSYGQ NDAKPPAVFT RISHYRPWIN KVLKQNKA | |
Greater than 90% as determined by SDS-PAGE. |
490628 | |
1 mg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |