missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7628-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000246882.1 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
30S ribosomal protein S2 Protein | |
Research Use Only | |
rpsB | |
Recombinant Protein | |
E. coli | |
MATVSMRDML KAGVHFGHQT RYWNPKMKPF IFGARNKVHI INLEKTVPMF NEALAELNKI ASRKGKILFV GTKRAASEAV KDAALSCDQF FVNHRWLGGM LTNWKTVRQS IKRLKDLETQ SQDGTFDKLT KKEALMRTRE LEKLENSLGG IKDMGGLPDA LFVIDADHEH IAIKEANNLG IPVFAIVDTN SDPDGVDFVI PGNDDAIRAV TLYLGAVAAT VREGRSQDLA SQAEESFVEA E | |
Greater than 90% as determined by SDS-PAGE. |