missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant E. coli Acetate kinase His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
0.5 mg
Unit Size:
0.10mg
0.5mg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-400 of E.coli Acetate kinase The Recombinant E. coli Acetate kinase Protein is derived from E. coli. The Recombinant E. coli Acetate kinase Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT |
| Molecular Weight (g/mol) | TMW: 45.7kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Common Name | Acetate kinase |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction