missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Cell division protein ZipA Protein

A DNA sequence encoding the Escherichia coli (strain K12) Cell division protein ZipA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6957-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16036890

  • 436.00 € / 10µg
Estimated Shipment: 05-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_416907.1
Cell division protein ZipA Protein
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
946869
Greater than 90% as determined by SDS-PAGE.
Research Use Only
zipA
Recombinant Protein
E. coli
MMQDLRLILI IVGAIAIIAL LVHGFWTSRK ERSSMFRDRP LKRMKSKRDD DSYDEDVEDD EGVGEVRVHR VNHAPANAQE HEAARPSPQH QYQPPYASAQ PRQPVQQPPE AQVPPQHAPH PAQPVQQPAY QPQPEQPLQQ PVSPQVAPAP QPVHSAPQPA QQAFQPAEPV AAPQPEPVAE PAPVMDKPKR KEAVIIMNVA AHHGSELNGE LLLNSIQQAG FIFGDMNIYH RHLSPDGSGP ALFSLANMVK PGTFDPEMKD FTTPGVTIFM QVPSYGDELQ NFKLMLQSAQ HIADEVGGVV LDDQRRMMTP QKLREYQDII REVKDANA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers