missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant E. coli IdHA His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.5mg
100µg
Specifications
Specifications
| Concentration | 1 mg/ml |
| For Use With (Application) | PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH8.0), 10% glycerol, 100 mM NaCl |
| Gene ID (Entrez) | 946315 |
| Molecular Weight (g/mol) | 39.1 kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Gene Alias | EC 1.1.1.28, ECK1377, hslF, hslI, htpH |
| Gene Symbol | ldhA |
| Purity or Quality Grade | >95% by SDS-PAGE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction