missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant E. coli IdHA His Protein

Product Code. p-200055036 Shop All Bio Techne Products
Click to view available options
Quantity:
0.5 mg
100 μg
Unit Size:
0.5mg
100µg
This item is not returnable. View return policy

Product Code. 18415211

Brand: Novus Biologicals™ NBP1988980.5mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/ml
For Use With (Application) PAGE
Formulation 20 mM Tris-HCl buffer (pH8.0), 10% glycerol, 100 mM NaCl
Gene ID (Entrez) 946315
Molecular Weight (g/mol) 39.1 kDa
Quantity 0.5 mg
Storage Requirements Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Gene Alias EC 1.1.1.28, ECK1377, hslF, hslI, htpH
Gene Symbol ldhA
Purity or Quality Grade >95% by SDS-PAGE
Protein IdHA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.