missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli insE1 Protein
A DNA sequence encoding the Escherichia coli (strain K12) insE1, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10112-ma-100ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_061380.1 | |
insE1 Protein | |
Research Use Only | |
insE1 | |
Recombinant Protein | |
Mammal | |
MTKTVSTSKK PRKQHSPEFR SEALKLAERI GVTAAARELS LYESQLYNWR SKQQNQQTSS ERELEMSTEI ARLKRQLAER DEELAILQKA ATYFAKRLK | |
Greater than 90% as determined by SDS-PAGE. |
1263530 | |
100 μg | |
Not determined. | |
Full Length | |
E. coli | |
Flag | |
Tris-based buffer, 50% glycerol |