missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli insE1 Protein

A DNA sequence encoding the Escherichia coli (strain K12) insE1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10112-ma-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15932878

  • 605.00 € / 20µg
Estimated Shipment: 19-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_061380.1
insE1 Protein
20 μg
Not determined.
Full Length
E. coli
Flag
Tris-based buffer, 50% glycerol
1263530
Greater than 90% as determined by SDS-PAGE.
Research Use Only
insE1
Recombinant Protein
Mammal
MTKTVSTSKK PRKQHSPEFR SEALKLAERI GVTAAARELS LYESQLYNWR SKQQNQQTSS ERELEMSTEI ARLKRQLAER DEELAILQKA ATYFAKRLK
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers