missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Outer membrane protein 1

A DNA sequence encoding the Escherichia coli O157:H7 Outer membrane protein A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7322-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048190

  • 940.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_309068.1
Outer membrane protein 1
Research Use Only
ompA
Recombinant Protein
E. coli
WTNNIGDAHT IGTRPDNGML SLGVSYRFGQ GEAAPVVAPA PAPAPEVQTK HFTLKSDVLF NFNKATLKPE GQAALDQLYS QLSNLDPKDG SVVVLGYTDR IGSDAYNQGL SERRAQSVVD YLISKGIPAD KISARGMGES NPVTGNTCDN VKQRAALIDC LAPDRRVEIE VKGIKDVVTQ PQA
Greater than 90% as determined by SDS-PAGE.
917123
100 μg
Not determined.
Partial (see sequence information for more details).
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers