Learn More
enQuireBio™ Recombinant hepatitis Capsid Protein
A DNA sequence encoding the Hepatitis E virus genotype 3 (isolate Human/United States/US2) (HEV-3) Capsid protein, was expressed in the hosts and tags indicated. option, abov
Brand: enQuireBio™ QP8294-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
hepatitis Capsid Protein | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Hepatitis | |
His | |
Tris-based buffer, 50% glycerol |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
ORF2 | |
Recombinant Protein | |
Yeast | |
KLYTSVENAQ QDKGITIPHD IDLGDSRVVI QDYDNQHEQD RPTPSPAPSR PFSVLRANDV LWLSLTAAEY DQTTYGSSTN PMYVSDTVTL VNVATGAMS- QAVARSLDWS KVTLDGRPLT TIQQYSKTFY VLPLRGKLSF WEAGTTKAGY PYNYNTTASD QILIENAAGH RVAISTYTTS LGAGPTSISA VGVLAPHSAL AVLEDTIDYP ARAHTFDDFC PECRTLGLQG CAFQSTIAEL QRLKMKVGKT RES |